Learn More
Nickel(II) sulfate hexahydrate, 99.97% min (metals basis), Thermo Scientific Chemicals
£64.62 - £283.41
Chemical Identifiers
CAS | 10101-97-0 |
---|---|
Molecular Formula | H12NiO10S |
Molecular Weight (g/mol) | 262.839 |
MDL Number | MFCD00149813 |
InChI Key | RRIWRJBSCGCBID-UHFFFAOYSA-L |
Synonym | nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 |
PubChem CID | 5284429 |
ChEBI | CHEBI:53437 |
IUPAC Name | nickel(2+);sulfate;hexahydrate |
SMILES | O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11498650
|
Thermo Scientific Alfa Aesar
053130.18 |
50 g |
£64.62
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488650
|
Thermo Scientific Alfa Aesar
053130.30 |
250 g |
£283.41
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nickel(II) sulfate hexahydrate is mainly used in Ni plating, as mordant in dyeing and in printing textiles, as a blackening agent for zinc and brass and in the fabrication of organic nickel salts. It is used as an effective catalyst for gas-phase dehydration of glycerol to acrolein. Nickel sulfate hexahydrate crystal is an efficient ultraviolet filter material. Columns used in biochemistry and molecular biology can be regenerated with nickel sulfate. It is used to prepare nickel carbonate, a precursor to pigments and nickel-based catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10101-97-0 | |
262.839 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
5284429 | |
nickel(2+);sulfate;hexahydrate |
H12NiO10S | |
MFCD00149813 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
CHEBI:53437 | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Specifications
10101-97-0 | |
Lump | |
H12NiO10S | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
nickel(2+);sulfate;hexahydrate | |
5284429 | |
262.86 (154.77 Anhydrous) | |
2.07 g/cm3 at 20°C |
Green | |
99.97% min. (metals basis) | |
MFCD00149813 | |
Very soluble in water,ethanol and ammonia. | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] | |
262.839 | |
CHEBI:53437 | |
Odorless | |
Nickel(II) sulfate hexahydrate |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
H302-H315-H317-H319-H332-H334-H335-H341-H350i-H360D-H372
missing translation for 'dotInformation' : DOT Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 232-104-9
missing translation for 'rtecsNumber' : QR9600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Nickel(II) sulfate hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Nickel(II) sulfate hexahydrate
RUO – Research Use Only