missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(Ii) Sulfate Hexahydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | NiSO4 · 6H2O |
UN Number | UN3077 |
Formula Weight | 262.85 |
Percent Purity | 99% |
Grade | ACS Reagent |
Density | Vapor Density: 2.07 (vs air) |
Chemical Name or Material | Nickel(Ii) Sulfate Hexahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'