missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) sulfate hexahydrate, Puriss. p.a., ACS Reagent, 99-102.0%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, 99-102.0%
£32.37 - £116.24
Chemical Identifiers
CAS | 10101-97-0 |
---|---|
Molecular Formula | H12NiO10S |
Molecular Weight (g/mol) | 262.839 |
MDL Number | MFCD00149813 |
InChI Key | RRIWRJBSCGCBID-UHFFFAOYSA-L |
Synonym | nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 |
PubChem CID | 5284429 |
ChEBI | CHEBI:53437 |
IUPAC Name | nickel(2+);sulfate;hexahydrate |
SMILES | O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10101-97-0 | |
262.839 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
5284429 | |
nickel(2+);sulfate;hexahydrate |
H12NiO10S | |
MFCD00149813 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
CHEBI:53437 | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Specifications
10101-97-0 | |
NiSO4 · 6H2O | |
3077 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
nickel(2+);sulfate;hexahydrate | |
5284429 | |
262.85g/mol | |
ACS/puriss. p.a. | |
Nickel(Ii) Sulfate Hexahydrate |
H12NiO10S | |
MFCD00149813 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] | |
262.839 | |
CHEBI:53437 | |
99 to 102% | |
Vapor Density: 2.07 (vs air) |
Safety and Handling
P201-P261-P273-P280-P284-P304 + P340 + P312
H302 + H332-H315-H317-H334-H341-H350i-H360D-H372-H410
missing translation for 'einecsNumber' : 232-104-9
missing translation for 'rtecsNumber' : QR9600000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title