missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfate hydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

£76.08 - £225.12

Chemical Identifiers

CAS 15244-37-8
Molecular Formula NiO4S
Molecular Weight (g/mol) 154.749
MDL Number MFCD00011146
InChI Key LGQLOGILCSXPEA-UHFFFAOYSA-L
Synonym nickel sulfate, nickel sulphate, nickel ii sulfate, nickelous sulfate, nickel 2+ sulfate, nickel sulfate 1:1, niso4, nickel ii sulfate 1:1, nickel sulfate van, nickel sulfate niso4
PubChem CID 24586
ChEBI CHEBI:53001
IUPAC Name nickel(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Ni+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346947
View Documents
Thermo Scientific Alfa Aesar
010820.14
25 g
£76.08
25g
Please to purchase this item. Need a web account? Register with us today!
11356947
View Documents
Thermo Scientific Alfa Aesar
010820.22
100 g
£225.12
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nickel(II) sulfate hydrate is used in electroplating and as a chemical intermediate to produce other nickel compounds. It is also used in printing and in dyeing of textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15244-37-8
154.749
LGQLOGILCSXPEA-UHFFFAOYSA-L
24586
nickel(2+);sulfate
NiO4S
MFCD00011146
nickel sulfate, nickel sulphate, nickel ii sulfate, nickelous sulfate, nickel 2+ sulfate, nickel sulfate 1:1, niso4, nickel ii sulfate 1:1, nickel sulfate van, nickel sulfate niso4
CHEBI:53001
[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

15244-37-8
4.3 to 4.7
NiO4S
MFCD00011146
14,6517
Soluble in water (29.3g/100cc water at 0°C and 83.7g/100cc water at 100°C. Insoluble in alcohol,ether & acetone.
[O-]S(=O)(=O)[O-].[Ni+2]
154.749
CHEBI:53001
99.9985%
Odorless
(metals basis)
Green
Crystalline Powder
NiSO4·xH2O (x≈6)
UN3288
nickel sulfate, nickel sulphate, nickel ii sulfate, nickelous sulfate, nickel 2+ sulfate, nickel sulfate 1:1, niso4, nickel ii sulfate 1:1, nickel sulfate van, nickel sulfate niso4
LGQLOGILCSXPEA-UHFFFAOYSA-L
nickel(2+);sulfate
24586
154.77 (Anhydrous)
Puratronic™
2.070 g/mL
Nickel(II) sulfate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c

H302+H332-H315-H317-H319-H334-H341-H350i-H360D-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 232-104-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only