Learn More
Nickel Iron Copper Molybdenum foil, 0.125mm (0.005 in.) thick, annealed (Mumetal™), Thermo Scientific Chemicals
Magnetic shielding alloy
£88.70 - £234.00
Specifications
Chemical Name or Material | Nickel Iron Copper Molybdenum foil |
---|---|
Name Note | 0.125mm (0.005 in.) thick, Annealed (Mumetal™) |
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Fe:Cu:Mo; 77:14:5:4 wt% |
MDL Number | MFCD02684505 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11355677
|
Thermo Scientific Alfa Aesar
043399.FL |
75 x 75 mm |
£88.70
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365677
|
Thermo Scientific Alfa Aesar
043399.GT |
150 x 150 mm |
£234.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nickel Iron Copper Molybdenum foil is used as a magnetic shielding alloy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel Iron Copper Molybdenum foil | |
Ni:Fe:Cu:Mo; 77:14:5:4 wt% | |
Insoluble in water. |
0.125mm (0.005 in.) thick, Annealed (Mumetal™) | |
MFCD02684505 | |
Foil |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Nickel Iron Copper Molybdenum foil, 0.125mm (0.005in) thick, annealed
Signal Word
- Danger
Hazard Category
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Nickel
RUO – Research Use Only