missing translation for 'onlineSavingsMsg'
Learn More
Nickel nitrate, Matrix Modifier Solution, Specpure™, Thermo Scientific Chemicals Product Code.: 15466295

Nickel nitrate, Matrix Modifier Solution, Specpure™, Thermo Scientific Chemicals

Product Code. 15466295
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 15466295

Brand: Thermo Scientific Alfa Aesar 039043.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7697-37-2, 13478-00-7, 7732-18-5
Color Green
Physical Form Liquid
Quantity 100 mL
Molecular Formula HNO3
MDL Number MFCD00011349
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Concentration or Composition (by Analyte or Components) Nitric acid: 2.0%; Nickel(II) nitrate hexahydrate: 1.0%; Water: 97.0%
Appearance Green liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Nickel nitrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1521708 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number039043.AE
Lot Number1481438 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number039043.AE
Lot Number1405655 Certificate TypeCertificate of Analysis Date16/10/2023 Catalog Number039043.AE
Lot Number1359913 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number039043.AE
Lot Number1359911 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number039043.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel nitrate, Matrix Modifier Solution, Specpure™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.