missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nickel/Plastic Bosshead

For use with rods with a maximum diameter of 16mm.

Brand:  Fisherbrand™ 2004

Product Code. 11755386


Description

Description

  • Supplied individually
  • With nickel plated steel screw

Specifications

Specifications

1 Ea.
Nickel, Plastic
For Rods up to 16mm dia.
Bosshead Clamp
Nickel plated brass screw Fitted blue Plastic headed screw
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers