Learn More
R&L Enterprises™ 3-Prong Clamps
Safely and securely holds articles in place in the laboratory. R&L Enterprises™ 3-Prong Clamps are manufactured from diecast aluminum alloy, actuated by nickel plated brass pummels and plastic headed nickel plated steel twin screw pivoting on a stainless steel pin.
Brand:  R&L Enterprises™ 1020
 View more versions of this product
Product Code. 10527514
  • £29.40 / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Jaws covered in chemically resistant plastic
  • For use with articles up to 90mm in diameter
  • Designed to hold articles within the jaws
  • Not recommended to hold articles by the points
  • Available with or without spring-loaded jaws
Specifications
Specifications
1 Ea.
Nickel, Plastic
Not Spring Loaded
3-Prong Clamp
90 mm