missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Nickel-plated Brass Luer Fittings
£31.80 - £37.30
Specifications
Autoclavable | Autoclavable |
---|---|
For Use With (Application) | Harsh chemical environments |
Packaging Quantity | 1 Pack |
Product Type | Luer Fitting |
Product Code | Brand | For Use With | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Includes | Price | Quantity & Availability | |||||
15392132
|
Cole-Parmer™
41507-62 |
Male Luer Lock | 1/16 in. Hose Barb |
£31.80
Each |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
15242146
|
Cole-Parmer™
41507-70 |
Male Luer Lock | 10-32 UNF Thread |
£37.30
Each |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
15312142
|
Cole-Parmer™
41507-64 |
Female luer lock | 1/16 in. Hose Barb | N/A | ||||||
Specifications
Autoclavable | |
Harsh chemical environments | |
1 Pack | |
Luer Fitting |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Nickel-plated Brass Luer Fittings