Learn More
Fisherbrand™ Hoffman Clips
For use with tubing up to 15mm in the laboratory.
Brand:  Fisherbrand™ 9503
Product Code. 12744396
  • £125.00 / Pack of 10
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With clamping screw hinges
  • Manufactured from plated nickel
  • 10 per pack
Specifications
Specifications
10/Pk.
Nickel-plated
For use with 15mm O.D. tubing
Clip Rexaloy
Clamping screw hinges