Learn More
Nickel plating solution, electroless, Thermo Scientific Chemicals
For Plating On Silicon, Germanium, Gallium Arsenide
£20.80 - £76.20
Specifications
Chemical Name or Material | Nickel plating solution |
---|---|
Physical Form | Liquid |
Color | Green |
MDL Number | MFCD00011137 |
Odor | Ammonia-like |
Description
For plating on silicon, germanium, gallium arsenide, cadmium sulfide, gold, nickel, iron and Kovar
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel plating solution | |
Green | |
Ammonia-like |
Liquid | |
MFCD00011137 |
Safety and Handling
H334-H350-H360-H372-H317-H341-H302-H332-H411-H401: May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. May cause an allergic skin reaction. Suspected of causing genetic defects. Harmful if swallowed. Harmful if inhaled. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
RUO – Research Use Only