Learn More
Nickel plating solution, semi-bright finish, Thermo Scientific Chemicals
£24.42 - £54.77
Specifications
Chemical Name or Material | Nickel plating solution |
---|---|
Color | Green |
Name Note | Semi-bright finish |
MDL Number | MFCD00011137 |
UN Number | UN3287 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11399687
|
Thermo Scientific Alfa Aesar
042026.AK |
250 mL |
£24.42
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389687
|
Thermo Scientific Alfa Aesar
042026.K2 |
1 L |
£54.77
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel plating solution | |
Semi-bright finish | |
UN3287 |
Green | |
MFCD00011137 | |
Liquid |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H302-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
H302+H332-H315-H317-H319-H334-H341-H350i-H360FD-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC LIQUID, INORGANIC, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only