Learn More
Nickel powder, -150+200 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
£37.59 - £350.75
Chemical Identifiers
CAS | 7440-02-0 |
---|---|
Molecular Formula | Ni |
Molecular Weight (g/mol) | 58.69 |
MDL Number | MFCD00011137 MFCD06798735 |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
PubChem CID | 935 |
ChEBI | CHEBI:28112 |
IUPAC Name | nickel |
SMILES | [Ni] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429980
|
Thermo Scientific Alfa Aesar
044739.22 |
100 g |
£37.59
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439980
|
Thermo Scientific Alfa Aesar
044739.36 |
500 g |
£110.55
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449980
|
Thermo Scientific Alfa Aesar
044739.A3 |
2 kg |
£350.75
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Specifications
Silver to Gray | |
Silver gray powder | |
99.8% (Metals basis) | |
MFCD00011137 MFCD06798735 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
-150+200 Mesh | |
Powder |
7440-02-0 | |
8.908g/cm3 at 20°C | |
Ni | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Odorless | |
Nickel |
Safety and Handling
Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
missing translation for 'einecsNumber' : 231-111-4
missing translation for 'rtecsNumber' : QR5950000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Nickel powder
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Nickel
RUO – Research Use Only