missing translation for 'onlineSavingsMsg'
Learn More

Nickel standard solution, for AAS, 1 mg/ml Ni in 2% HNO3, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 196151000


 View more versions of this product

Product Code. 10226780

  • £17.09 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nickel standard solution
1 mg/ml Ni in 2% HNO3
Plastic bottle
Green
2.1, 97.1
MFCD00011137 MFCD06798735
[Ni]
58.69
CHEBI:28112
AAS
1.032
Crystals, Crystalline Powder and/or Chunks
990 to 1010μg/mL Ni in 2 to 5% HNO3 (20°C)
7440-02-0, 13478-00-7, 7697-37-2, 7732-18-5
100 mL
1.9, 96.9
Ni
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
935
58.69
1.0320g/mL
Soluble in water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Nickel standard solution, 1 mg/ml Ni in 2-5% HNO3

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only