missing translation for 'onlineSavingsMsg'
Learn More
Nickel Thinfoil, 0.0025mm (0.0001in) thick, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11378638

Nickel Thinfoil, 0.0025mm (0.0001in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11378638
100 x 100 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
This item is not returnable. View return policy

Product Code. 11378638

Brand: Thermo Scientific Alfa Aesar 040714.GH

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 100 x 100 mm
Percent Purity 99.95%
Odor Odorless
Weight ≈0.014g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Nickel Thinfoil, 0.0025mm (0.0001 in.) thick, Not certified pinhole free

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS28J030 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog Number040714.FI,040714,040714.FF,040714.GH
Lot NumberR10I052 Certificate TypeCertificate of Analysis Date13/06/2022 Catalog Number040714.FI,040714.FF,040714.GH
Lot NumberR24I009 Certificate TypeCertificate of Analysis Date01/06/2022 Catalog Number040714.FI,040714.FF,040714.GH
Lot NumberY05H010 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040714.FI,040714.FF,040714.GH
Lot NumberD22T035 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number040714.FI,040714.FF,040714.GH

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel Thinfoil, 0.0025mm (0.0001in) thick, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.