missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Nickel tungsten oxide is used as photo catalysts, sensors and humidity dielectric resonators. It is considered as a new cathode material for asymmetric super capacitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Powder |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Formula Weight | 306.53 |
Percent Purity | 99.9% |
Odor | Odorless |
Assay | (metals basis excluding Co), Co <100ppm |
Chemical Name or Material | Nickel tungsten oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'