missing translation for 'onlineSavingsMsg'
Learn More
Nickel wire, 1.0mm (0.04in) dia, annealed, 99.98% (metals basis), Thermo Scientific Chemicals Product Code.: 11364808

Nickel wire, 1.0mm (0.04in) dia, annealed, 99.98% (metals basis), Thermo Scientific Chemicals

Product Code. 11364808
50 m
Click to view available options
Quantity:
10 m
2 m
50 m
This item is not returnable. View return policy

Product Code. 11364808

Brand: Thermo Scientific Alfa Aesar 043133.H2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 7.0g/m, Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Assay Percent Range 99.98%, metal basis
Quantity 50 m
Odor Odorless
Form Wire
Chemical Name or Material Nickel

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel wire, 1.0mm (0.04in) dia, annealed, 99.98% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.