missing translation for 'onlineSavingsMsg'
Learn More
Nickel wire, 2.0mm (0.08in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals Product Code.: 11301468

Nickel wire, 2.0mm (0.08in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11301468
25 cm
Click to view available options
Quantity:
5 cm
25 cm
100 cm
This item is not returnable. View return policy

Product Code. 11301468

Brand: Thermo Scientific Alfa Aesar 010928.BU

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx.28.0g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Color Silver to Gray
Appearance Silver gray wire
Assay Percent Range 99.995% (Metals basis)
Quantity 25 cm
Odor Odorless
Form Wire
Chemical Name or Material Nickel

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX14K022 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number010928.BQ,010928.BU,010928.BY
Lot NumberG27Y049 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010928.BQ,010928.BU,010928.BY
Lot NumberQ18C002 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010928.BQ,010928.BU,010928.BY
Lot NumberR29A036 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010928.BQ,010928.BU,010928.BY
Lot NumberG07Q22 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010928.BQ,010928.BU,010928.BY
compliance-icons
Product Identifier
  • Nickel wire
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : Nickel

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel wire, 2.0mm (0.08in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.