missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinamide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
1000 g
Description
Nicotinamide is a building block (active component) of coenzymes nicotinamide adenine dinucleotide (NAD) and nicotinamide adenine dinucleotide phopshate (NADP). It may be used as a substrate of nicotinamide phosphoribosyltransferase (NAMPT, EC 2.4.2.12), the rate-limiting enzyme for NAD(+) biosynthesis and an important signaling molecule. It is an amide derivative of vitamin B3 and a PARP inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nicotinamide is a building block (active component) of coenzymes nicotinamide adenine dinucleotide (NAD) and nicotinamide adenine dinucleotide phopshate (NADP). It may be used as a substrate of nicotinamide phosphoribosyltransferase (NAMPT, EC 2.4.2.12), the rate-limiting enzyme for NAD(+) biosynthesis and an important signaling molecule. It is an amide derivative of vitamin B3 and a PARP inhibitor.
Solubility
Soluble in water, ethanol , glycerol, DMSO , methanol, butanol and chloroform. Slightly soluble in ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Nicotinamide is a building block (active component) of coenzymes nicotinamide adenine dinucleotide (NAD) and nicotinamide adenine dinucleotide phopshate (NADP). It may be used as a substrate of nicotinamide phosphoribosyltransferase (NAMPT, EC 2.4.2.12), the rate-limiting enzyme for NAD(+) biosynthesis and an important signaling molecule. It is an amide derivative of vitamin B3 and a PARP inhibitor.
Solubility
Soluble in water, ethanol , glycerol, DMSO , methanol, butanol and chloroform. Slightly soluble in ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 128°C to 132°C |
Flash Point | 182°C (360°F) |
Assay Percent Range | 99% |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 383619 |
Merck Index | 14,6523 |
Formula Weight | 122.13 |
Percent Purity | 99% |
Chemical Name or Material | Nicotinamide |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Nicotinamide
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : nicotinamide
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'