missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Nicotinamide Adenine Dinucleotide, MP Biomedicals™
Click to view available options
Quantity:
5 g
25 g
Description
- Serves as a coenzyme of the dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols
- Usually acts as a hydrogen acceptor, forming NADH, which then serves as a hydrogen donor in the respiratory chain
- Acts as a coenzyme for hydride-transfer enzymes
- Used as a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins
- Recently reported to be a P2Y11 agonist
Specifications
Specifications
pH | 2 to 4 (1% aq. soln.) |
Quantity | 25 g |
Solubility Information | Soluble in water (200mg/mL) |
Formula Weight | 663.40 (anhydrous) |
Percent Purity | ≥93% |
Physical Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction