missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate, 97+%, Thermo Scientific Chemicals
£56.90 - £583.00
Chemical Identifiers
CAS | 698999-85-8 |
---|---|
Molecular Formula | C21H27N7NaO17P3 |
Molecular Weight (g/mol) | 765.39 |
MDL Number | MFCD00036973 |
InChI Key | JNUMDLCHLVUHFS-QYZPTAICSA-M |
Synonym | nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium |
PubChem CID | 131845262 |
SMILES | [Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10287303
|
Thermo Scientific Acros
227571000 |
100 mg | Glass bottle |
£56.90
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10144570
|
Thermo Scientific Acros
227575000 |
500 mg | Glass bottle |
£177.00
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11906821
|
Thermo Scientific Acros
227570025 |
2.5 g | Glass bottle |
£583.00
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
698999-85-8 | |
765.39 | |
JNUMDLCHLVUHFS-QYZPTAICSA-M | |
131845262 |
C21H27N7NaO17P3 | |
MFCD00036973 | |
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium | |
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
Specifications
175.0°C to 178.0°C | |
White to Yellow | |
determination,based on dry weight) | |
MFCD00036973 | |
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium | |
JNUMDLCHLVUHFS-QYZPTAICSA-M | |
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium | |
131845262 | |
≥97% | |
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate |
698999-85-8 | |
Powder | |
C21H27N7NaO17P3 | |
14,6348 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,less soluble in ethanol,practically insoluble in ether and ethyl acetate | |
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O | |
765.39 | |
765.39 | |
Authentic |
Safety and Handling
missing translation for 'einecsNumber' : 214-664-6
RUO – Research Use Only