Learn More
β-Nicotinamide mononucleotide, MedChemExpress
MedChemExpress β-nicotinamide mononucleotide (β-NM) is a product of the nicotinamide phosphoribosyltransferase (NAMPT) reaction and a key NAD+ intermediate. The pharmacological activities of β-nicotinamide mononucleotide include its role in cellular biochemical functions, cardioprotection, diabetes, Alzheimer's disease, and complications associated with obesity.
Brand:  Medchem Express HY-F0004
Encompass_Preferred
 View more versions of this product
Product Code. 16468926
  • £261.36 / 100mg
Explore more special offers
Description
Description
β-nicotinamide mononucleotide (β-NM) is a product of the nicotinamide phosphoribosyltransferase (NAMPT) reaction and a key NAD+ intermediate. The pharmacological activities of β-nicotinamide mononucleotide include its role in cellular biochemical functions, cardioprotection, diabetes, Alzheimer's disease, and complications associated with obesity.
Specifications
Specifications
β-Nicotinamide mononucleotide
White
C11H15N2O8P
β-NM NMN
NC(C1=C[N+]([C@@H]2O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]2O)=CC=C1)=O
334.22
Research
1094-61-7
Research
100 mg
H2O : 83.33 mg/mL (249.33 mM; Need ultrasonic)
334.22
98.0%
Solid