missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide, Tocris Bioscience™

Code produit. 18782534
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

PARP-1 inhibitor

Nicotinamide is an anti-inflammatory agent. Inhibitor of poly(ADP-ribose) polymerase (PARP-1) enzymes. NAD+ precursor. Promotes differentiation of mesenchymal stem cells to insulin producing cells when used in combination with growth factors and high glucose concentration. Nicotinamide also acts as a SIRT1 inhibitor and promotes the expansion of hematopoietic progenitor cells

For more information about how Nicotinamide may be used, see our protocol:

3D Culture of Lung Alveolar Cells

Spécification

CAS 98-92-0
Molecular Formula C6H6N2O
Quantity 50 mg
Synonym nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
SMILES C1=CC(=CN=C1)C(=O)N
IUPAC Name pyridine-3-carboxamide
Molecular Weight (g/mol) 122.127
PubChem CID 936
ChEBI CHEBI:17154
Formula Weight 122.12
Percent Purity >99%
Chemical Name or Material Nicotinamide
Afficher plus Afficher moins
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'