missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic acid, 99.5%

Product Code. 10081320 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10081320

Brand: Thermo Scientific Acros 128295000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-67-6 | C6H5NO2 | 123.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
Synonym nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
PubChem CID 938
ChEBI CHEBI:15940
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point 236.0°C to 239.0°C
Color White
Loss on Drying 1% max. (105°C, 1 hr.)
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Plastic bottle
Quantity 500 g
Beilstein 22,38
Merck Index 15,661
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: slightly soluble in ethanol (10g/L at 20°C),eth,er,soluble in propylene glycol,in alkali hydroxides,and carbonates,freely soluble in boiling water or boiling ethanol
Formula Weight 123.11
Percent Purity 99.5%
Physical Form Powder
Chemical Name or Material Nicotinic acid, 99.5%
Show More Show Less
compliance-icons
Product Identifier
  • Nicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.