missing translation for 'onlineSavingsMsg'
Learn More

Nigrosin, alcohol soluble, Thermo Scientific Chemicals

Product Code. 15463809
Click to view available options
Quantity:
25 g
100 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11099-03-9
Molecular Formula C8H19ClN2O
Molecular Weight (g/mol) 194.703
MDL Number MFCD00149005
InChI Key GBPWTMZRCAESKB-UHFFFAOYSA-N
Synonym Solvent Black 5; C.I. 50415
PubChem CID 19828004
IUPAC Name 2-amino-N,N-dipropylacetamide;hydrochloride
SMILES CCCN(CCC)C(=O)CN.Cl

Specifications

Solubility Information Soluble in alcohol
Color Black
Quantity 100 g
Physical Form Powder
Chemical Name or Material Nigrosin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'