missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nimorazole, Honeywell Fluka™
Click to view available options
Quantity:
10 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Solid |
CAS | 6506-37-2 |
Molecular Formula | C9H14N4O3 |
InChI Key | MDJFHRLTPRPZLY-UHFFFAOYSA-N |
SMILES | [O-][N+](=O)C1=CN=CN1CCN1CCOCC1 |
IUPAC Name | 4-[2-(5-nitro-1H-imidazol-1-yl)ethyl]morpholine |
Molecular Weight (g/mol) | 226.24 |
Grade | Analytical Standard |
Packaging | Glass Bottle |
Quantity | 10 mg |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'