missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nimotuzumab Recombinant Human Monoclonal Antibody (h-R3 (Nimotuzumab))
GREENER_CHOICE

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA548049

Product Code. 17971271

  • £464.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specificity: Binds to human EGFR domain III.

Nimotuzumab is a humanized therapeutic monoclonal antibody against epidermal growth factor receptor (EGFR).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nimotuzumab
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
Human
Protein A
RUO
Human
Antibody
IgG1 κ
Flow Cytometry, Inhibition Assays, Immunohistochemistry
h-R3 (Nimotuzumab)
Unconjugated
hR3; Theraloc
This antibody we generated by humanisation of the murine antibody R3, raised against EGFR purified from placenta membranes and A431 tumors.
200 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.