Learn More
Ninhydrin, 99%, Thermo Scientific Chemicals
£19.20 - £233.00
Chemical Identifiers
CAS | 485-47-2 |
---|---|
Molecular Formula | C9H6O4 |
Molecular Weight (g/mol) | 178.143 |
MDL Number | MFCD00003791 |
InChI Key | FEMOMIGRRWSMCU-UHFFFAOYSA-N |
Synonym | ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione |
PubChem CID | 10236 |
ChEBI | CHEBI:86374 |
IUPAC Name | 2,2-dihydroxyindene-1,3-dione |
SMILES | C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458603
|
Thermo Scientific Alfa Aesar
A10409.09 |
10 g |
£19.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488730
|
Thermo Scientific Alfa Aesar
A10409.14 |
25 g |
£39.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468603
|
Thermo Scientific Alfa Aesar
A10409.18 |
50 g |
£61.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478603
|
Thermo Scientific Alfa Aesar
A10409.30 |
250 g |
£233.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNinhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.
Solubility
Soluble in water, acetone, ethanol, and methanol. Slightly soluble in chloroform and ether.
Notes
Light Sensitive. Incompatible with strong oxidizing agents, amines, alkalis and strong bases.
Chemical Identifiers
485-47-2 | |
178.143 | |
FEMOMIGRRWSMCU-UHFFFAOYSA-N | |
10236 | |
2,2-dihydroxyindene-1,3-dione |
C9H6O4 | |
MFCD00003791 | |
ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione | |
CHEBI:86374 | |
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O |
Specifications
485-47-2 | |
99% | |
MFCD00003791 | |
Light sensitive | |
ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione | |
FEMOMIGRRWSMCU-UHFFFAOYSA-N | |
2,2-dihydroxyindene-1,3-dione | |
10236 | |
178.14 | |
Powder |
251°C (decomposition) | |
C9H6O4 | |
1910963 | |
14,6554 | |
Soluble in water,acetone,ethanol,and methanol. Slightly soluble in chloroform and ether. | |
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O | |
178.143 | |
CHEBI:86374 | |
99% | |
Ninhydrin |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 207-618-1
missing translation for 'rtecsNumber' : NK5425000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only