missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ninhydrin, Reagent ACS

Product Code. 10287183
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00003791
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym 1, 2, 3-Indantrione, 1, 2, 3-Triketohydrindene, 2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione, 1H-Indene-1, 2, 3-trione
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Specifications

Melting Point 250.0°C
Fieser 01,732; 04,356
Merck Index 15, 6640
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, slightly soluble in ether and chloroform
Identification Passes Test
Infrared Spectrum Authentic
Formula Weight 178.14
Grade ACS Reagent
Packaging Glass bottle
Sensitivity (to amino acids) Passes Test
Density 0.81
Color Brown/White to White
Quantity 1 kg
Physical Form Crystals
Chemical Name or Material Ninhydrin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'