Learn More
Thermo Scientific Chemicals Ninhydrin, Reagent ACS
£34.60 - £962.00
Chemical Identifiers
CAS | 485-47-2 |
---|---|
Molecular Formula | C9H6O4 |
Molecular Weight (g/mol) | 178.143 |
MDL Number | MFCD00003791 |
InChI Key | FEMOMIGRRWSMCU-UHFFFAOYSA-N |
Synonym | 1, 2, 3-Indantrione, 1, 2, 3-Triketohydrindene, 2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione, 1H-Indene-1, 2, 3-trione |
PubChem CID | 10236 |
ChEBI | CHEBI:86374 |
IUPAC Name | 2,2-dihydroxyindene-1,3-dione |
SMILES | C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10055780
|
Thermo Scientific Chemicals
415720100 |
10 g | Glass bottle |
£34.60
10g |
In Stock
Log in to see stock. |
||||
10404701
|
Thermo Scientific Chemicals
415720250 |
25 g | Glass Bottle |
£39.40
25g |
In Stock
Log in to see stock. |
||||
10334792
|
Thermo Scientific Chemicals
415721000 |
100 g | Glass bottle |
£109.00
100g |
In Stock
Log in to see stock. |
||||
10287183
|
Thermo Scientific Chemicals
415720010 |
1 kg | Glass bottle |
£962.00
1kg |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
485-47-2 | |
178.143 | |
FEMOMIGRRWSMCU-UHFFFAOYSA-N | |
10236 | |
2,2-dihydroxyindene-1,3-dione |
C9H6O4 | |
MFCD00003791 | |
1, 2, 3-Indantrione, 1, 2, 3-Triketohydrindene, 2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione, 1H-Indene-1, 2, 3-trione | |
CHEBI:86374 | |
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O |
Specifications
250.0°C | |
C9H6O4 | |
01,732; 04,356 | |
1, 2, 3-Indantrione, 1, 2, 3-Triketohydrindene, 2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione, 1H-Indene-1, 2, 3-trione | |
Passes Test | |
Authentic | |
2,2-dihydroxyindene-1,3-dione | |
10236 | |
178.14 | |
(to amino acids) Passes Test | |
Brown/White to White | |
Ninhydrin |
485-47-2 | |
MFCD00003791 | |
15, 6640 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, slightly soluble in ether and chloroform | |
FEMOMIGRRWSMCU-UHFFFAOYSA-N | |
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O | |
178.143 | |
CHEBI:86374 | |
ACS Reagent | |
0.81 | |
Crystals |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 207-618-1
missing translation for 'rtecsNumber' : NK5425000
missing translation for 'tsca' : TSCA
- Ninhydrin
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only