missing translation for 'onlineSavingsMsg'
Learn More

Niobium carbide, 99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11346518
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Niobium carbide is used in tool bits for cutting tools.  Zirconium carbide and niobium carbide can be used as refractory coatings in nuclear reactors. It is used to produce arc-welding rods and in corrosion-resistant steel. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-94-2
Molecular Formula CNb
Molecular Weight (g/mol) 104.92
MDL Number MFCD00016237
InChI Key AADPBXLVMHIYCT-UHFFFAOYSA-N
Synonym niobium carbide nbc, methane-niobium 1:1, methane niobium, niobium carbide trace metals basis 25g
PubChem CID 159431
IUPAC Name methane;niobium
SMILES [C-]#[Nb+]

Specifications

Melting Point 3500°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99+% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 104.91
Odor Odorless
Appearance Gray powder
Density 7.6
Chemical Name or Material Niobium carbide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'