missing translation for 'onlineSavingsMsg'
Learn More
Niobium carbide, 99+% (metals basis), Thermo Scientific Chemicals Product Code.: 11356518

Niobium carbide, 99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11356518
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11356518

Brand: Thermo Scientific Alfa Aesar 012147.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Niobium carbide is used in tool bits for cutting tools.  Zirconium carbide and niobium carbide can be used as refractory coatings in nuclear reactors. It is used to produce arc-welding rods and in corrosion-resistant steel. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-94-2
Molecular Formula CNb
Molecular Weight (g/mol) 104.92
MDL Number MFCD00016237
InChI Key AADPBXLVMHIYCT-UHFFFAOYSA-N
Synonym niobium carbide nbc, methane-niobium 1:1, methane niobium, niobium carbide trace metals basis 25g
PubChem CID 159431
IUPAC Name methane;niobium
SMILES [C-]#[Nb+]

Specifications

Melting Point 3500°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99+% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 104.91
Odor Odorless
Appearance Gray powder
Density 7.6 g/mL
Chemical Name or Material Niobium carbide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN21L021 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog Number012147.22,012147.14
Lot NumberN03L034 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog Number012147.22,012147.14
Lot NumberW14K004 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number012147.22,012147.14
Lot NumberU31J036 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number012147.22,012147.14
Lot NumberY03I050 Certificate TypeCertificate of Analysis Date09/11/2022 Catalog Number012147.22,012147.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium carbide, 99+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.