missing translation for 'onlineSavingsMsg'
Learn More
Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11371318

Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11371318
50 x 100 mm
Click to view available options
Quantity:
50 x 100 mm
100 x 200 mm
200 x 500 mm
This item is not returnable. View return policy

Product Code. 11371318

Brand: Thermo Scientific Alfa Aesar 010257.FY

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx.42.9g/50 x 100mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-03-1
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Color Silver to Gray
Appearance Silver gray foil
Assay Percent Range 99.80%
Quantity 50 x 100 mm
Odor Odorless
Form Foil
Chemical Name or Material Niobium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX17K048 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog Number010257.HQ,010257.FY,010257.GK
Lot NumberM20J013 Certificate TypeCertificate of Analysis Date01/02/2023 Catalog Number010257.HQ,010257.FY,010257.GK
Lot NumberK09P07 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010257.HQ,010257.FY,010257.GK
Lot NumberD10I14 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010257.HQ,010257.FY,010257.GK
Lot NumberK06S029 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010257.HQ,010257.FY,010257.GK

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.