missing translation for 'onlineSavingsMsg'
Learn More

Niobium ingot/button,≈25.4mm (1.0 in.) dia. x 11mm (0.45 in.) thick, 99.95% (metals basis)

Product Code. 15454945
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15454945

Brand: Thermo Scientific Alfa Aesar 045113.KF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-03-1 | Nb | 92.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Silver to Gray
CAS 3-1-7440
Quantity 1 Pc.
Odor Odorless
Weight ≈50g/piece
Form Pieces
Molecular Formula Nb
MDL Number MFCD00011126
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES [Nb]
IUPAC Name niobium
Molecular Weight (g/mol) 92.91
PubChem CID 23936
ChEBI CHEBI:33344
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Niobium ingot/button, ≈25.4mm (1.0 in.) dia. x 11mm (0.45 in.) thick
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.