missing translation for 'onlineSavingsMsg'
Learn More
Niobium(IV) selenide, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11382188

Niobium(IV) selenide, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11382188
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11382188

Brand: Thermo Scientific Alfa Aesar 013101.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a potential substance for thin film solar cells as back contact material. It is widely used in electricity brush & special bearings because of its conductivity and lubricity property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12034-77-4
Molecular Formula NbSe2
Molecular Weight (g/mol) 250.85
MDL Number MFCD00016228
InChI Key CXRFFSKFQFGBOT-UHFFFAOYSA-N
Synonym niobium selenide, columbium selenide, niobium diselenide, diselanylideneniobium, bis selanylidene niobium, niobium diselenide, crystal, niobium diselenide trace metals basis, niobium iv selenide trace metals basis 10gShow MoreShow Less
PubChem CID 82841
IUPAC Name bis(selanylidene)niobium
SMILES [Se]=[Nb]=[Se]

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3283
Solubility Information Insoluble in water.
Formula Weight 250.83
Percent Purity 99.8%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Niobium(IV) selenide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR17I039 Certificate TypeCertificate of Analysis Date02/08/2022 Catalog Number013101.18,013101.09
Lot NumberF30X012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013101.18,013101.09
Lot NumberG06P34 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013101.18,013101.09
Lot NumberJ28T004 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013101.18,013101.09
Lot NumberD07N34 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013101.18,013101.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium(IV) selenide, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.