missing translation for 'onlineSavingsMsg'
Learn More
Niobium powder, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11301348

Niobium powder, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11301348
100 g
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11301348

Brand: Thermo Scientific Alfa Aesar 010275.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-03-1
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

UN Number UN3089
Quantity 100 g
Percent Purity 99.8%
Mesh Size −325 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Niobium powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10198007 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog Number010255.36,010255.A3,010255.22,010275.22,010255.09
Lot Number10198006 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog Number010255.36,010255.A3,010255.22,010275.22,010255.09
Lot NumberX29H053 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010315.14,010275.22,010275.88,010275.14,010275.36
Lot NumberK08Q025 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010315.14,010275.22,010275.88,010275.14,010275.36
Lot NumberJ24E24 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010315.14,010275.22,010275.88,010275.14,010275.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium powder, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.