missing translation for 'onlineSavingsMsg'
Learn More

Niobium silicide, 99.85% (metals basis), Thermo Scientific Chemicals

£87.74 - £240.20

Chemical Identifiers

CAS 12034-80-9
Molecular Formula NbSi2
Molecular Weight (g/mol) 149.076
MDL Number MFCD00151537
InChI Key GXAAOBARRLPQRY-UHFFFAOYSA-N
Synonym niobium silicide, niobium disilicide, bis $l^ 2, columbium silicide,
PubChem CID 6336884
IUPAC Name bis($l^{2}-silanylidene)niobium
SMILES [Si]=[Nb]=[Si]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11372188
SDS View Documents
Thermo Scientific Alfa Aesar
013100.14
25 g
£87.74
25g
Please to purchase this item. Need a web account? Register with us today!
11362188
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
013100.22
100 g
£240.20
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Niobium silicide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

missing translation for 'einecsNumber' : 234-812-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium silicide, 99.85% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.