missing translation for 'onlineSavingsMsg'
Learn More

Niobium slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis excluding Ta)

Product Code. p-7058599
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11340068

Brand: Thermo Scientific Alfa Aesar 042845.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vapor deposition | CAS: 7440-03-1 | Nb | 92.91 g/mol

For vapor deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-1-7440
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Quantity 10 g
Odor Odorless
Weight ≈0.22g/slug
Form Slug
Percent Purity 99.95%
Assay (metals basis excluding Ta)
Chemical Name or Material Niobium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.