missing translation for 'onlineSavingsMsg'
Learn More
Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals Product Code.: 10398670

Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals

Product Code. 10398670
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10398670

Brand: Thermo Scientific 201690500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-12-7
Molecular Formula Cl5Nb
Molecular Weight (g/mol) 270.16
MDL Number MFCD00011127
InChI Key YHBDIEWMOMLKOO-UHFFFAOYSA-I
Synonym niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratremShow MoreShow Less
PubChem CID 24818
IUPAC Name pentachloroniobium
SMILES Cl[Nb](Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 254.0°C
Melting Point 204.7°C to 209.5°C
Color Yellow
CAS Min % 99.75
CAS Max % 100.0
Physical Form Fine Crystalline Powder
Quantity 50 g
Assay Percent Range 99.75% min
Linear Formula NbCl5
Fieser 14,214
Merck Index 15, 6643
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in hcl, soluble in ccl4, soluble in ethanol
Formula Weight 270.16
Percent Purity 99.8%
Packaging Glass bottle
Chemical Name or Material Niobium(V) chloride, 99.8%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463741 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number201690100,201690500
Lot NumberA0459974 Certificate TypeCertificate of Analysis Date07/02/2024 Catalog Number201690100,201690500
Lot NumberA0405600 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number201690100,201690500
Lot NumberA0417606 Certificate TypeCertificate of Analysis Date12/09/2022 Catalog Number201690100,201690500
Lot NumberA0437308 Certificate TypeCertificate of Analysis Date16/12/2021 Catalog Number201690100,201690500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.