missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) chloride, 99.9% (metals basis)

Product Code. 11317257
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11317257

Brand: Thermo Scientific Alfa Aesar 011548.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-12-7 | Cl5Nb | 270.156 g/mol

It is the precursor in the preparation of pure niobium, and ferroniobium. It is used as Lewis acid catalyst in the organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-12-7
Molecular Formula Cl5Nb
Molecular Weight (g/mol) 270.156
MDL Number MFCD00011127
InChI Key YHBDIEWMOMLKOO-UHFFFAOYSA-I
Synonym niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
PubChem CID 24818
IUPAC Name pentachloroniobium
SMILES Cl[Nb](Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 248.2°C
Melting Point 208.3°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in alcohol,hydrochloric acid,carbon tetrachloride and chloroform.
Formula Weight 270.2
Odor Acidic
Sensitivity Moisture sensitive
Appearance Yellow powder/flake/crystalline/beads
Density 2.74 g/cm3 at 20°C
Chemical Name or Material Niobium(V) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.