missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) chloride, 99.95% (metals basis), Thermo Scientific Chemicals

£47.54 - £185.93

Chemical Identifiers

CAS 10026-12-7
Molecular Formula Cl5Nb
Molecular Weight (g/mol) 270.156
MDL Number MFCD00011127
InChI Key YHBDIEWMOMLKOO-UHFFFAOYSA-I
Synonym niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
PubChem CID 24818
IUPAC Name pentachloroniobium
SMILES Cl[Nb](Cl)(Cl)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11308697
View Documents
Thermo Scientific Alfa Aesar
035795.09
10 g
£47.54
10g
Please to purchase this item. Need a web account? Register with us today!
11398687
View Documents
Thermo Scientific Alfa Aesar
035795.18
50 g
£185.93
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used to form mixed chloro-aryloxide compounds, such as [Nb(OC6H3-2,6-i-Pr2)2Cl3]2, which form pyridine or phosphine adducts with a distorted six-coordinate geometry. It is also used in the preparation of pure niobium, and as an intermediate, precursor to many other laboratory reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

10026-12-7
270.156
YHBDIEWMOMLKOO-UHFFFAOYSA-I
24818
Cl[Nb](Cl)(Cl)(Cl)Cl
Cl5Nb
MFCD00011127
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
pentachloroniobium

Specifications

248.2°C
10026-12-7
Cl5Nb
UN3260
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
YHBDIEWMOMLKOO-UHFFFAOYSA-I
pentachloroniobium
24818
99.95%
Moisture sensitive
(metals basis)
208.3°C
Crystalline
MFCD00011127
14,6557
Soluble in water,alcohol,HCl,and Carbon tetrachloride.
Cl[Nb](Cl)(Cl)(Cl)Cl
270.156
270.2
Acidic
2.74 g/mL
Niobium(V) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 233-059-8

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only