missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) ethoxide, 99.9% (metals basis)

Product Code. p-7046728
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11372867

Brand: Thermo Scientific Alfa Aesar L16031.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3236-82-6 | C10H25NbO5 | 318.21 g/mol

Niobium(V) ethoxide is used in nano-materials & thin films, catalysis & synthesis, hybrid & disperse materials, ferroelectric metals. It is mainly used for the sol-gel processing of materials containing niobium oxides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Niobium(V) ethoxide is used in nano-materials & thin films, catalysis & synthesis, hybrid & disperse materials, ferroelectric metals. It is mainly used for the sol-gel processing of materials containing niobium oxides

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. It is sensitive to moisture. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3236-82-6
Molecular Formula C10H25NbO5
Molecular Weight (g/mol) 318.21
MDL Number MFCD00015122
InChI Key ZTILUDNICMILKJ-UHFFFAOYSA-N
Synonym niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+
PubChem CID 160675
SMILES CCO[Nb](OCC)(OCC)(OCC)OCC

Specifications

Boiling Point 140°C to 142°C (0.1mmHg)
Melting Point 6°C
Quantity 1 g
UN Number UN2924
Beilstein 3678883
Flash Point 36°C (97°F)
Solubility Information Reacts with water.
IUPAC Name pentaethoxyniobium
Formula Weight 318.21
Percent Purity 99.9%
Refractive Index 1.516
Sensitivity Moisture sensitive
Density 1.258
Assay (metals basis)
Chemical Name or Material Niobium(V) ethoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.