missing translation for 'onlineSavingsMsg'
Learn More
Niobium(V) ethoxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10145900

Niobium(V) ethoxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10145900
5 g, Glass bottle
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10145900

Brand: Thermo Scientific Acros 316880050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3236-82-6
Molecular Formula C10H25NbO5
Molecular Weight (g/mol) 318.21
MDL Number MFCD00015122
InChI Key ZTILUDNICMILKJ-UHFFFAOYSA-N
Synonym niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+Show MoreShow Less
PubChem CID 160675
SMILES CCO[Nb](OCC)(OCC)(OCC)OCC

Specifications

Boiling Point 140.0°C to 142.0°C (0.1mmHg)
Melting Point 6.0°C
CAS Min % 99.9
CAS Max % 100.0
Physical Form Liquid
Quantity 5 g
Assay Percent Range 99.9%
Linear Formula Nb(OC2H5)5
Flash Point 36°C
Solubility Information Solubility in water: reacts
IUPAC Name pentaethoxyniobium
Formula Weight 318.21
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 1.268
Density 1.2680g/mL
Chemical Name or Material Niobium(V) ethoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462978 Certificate TypeCertificate of Analysis Date22/05/2024 Catalog Number316880050,316880500
Lot NumberA0444984 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number316880050,316880500
Lot NumberA0403206 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number316880050,316880500
Lot NumberA0439182 Certificate TypeCertificate of Analysis Date18/02/2022 Catalog Number316880050,316880500
Lot NumberA0423561 Certificate TypeCertificate of Analysis Date20/10/2020 Catalog Number316880050,316880500
compliance-icons compliance-icons
Product Identifier
  • Niobium(V) ethoxide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium(V) ethoxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.