missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

For Use As An Intermediate and in Electronics

£73.60 - £1088.42

Chemical Identifiers

CAS 1313-96-8
Molecular Formula Nb2O5
Molecular Weight (g/mol) 265.81
MDL Number MFCD00011128
InChI Key ZKATWMILCYLAPD-UHFFFAOYSA-N
Synonym niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
PubChem CID 123105
SMILES O=[Nb](=O)O[Nb](=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311149
View Documents
Thermo Scientific Alfa Aesar
011365.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11301149
View Documents
Thermo Scientific Alfa Aesar
011365.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11321149
View Documents
Thermo Scientific Alfa Aesar
011365.A3
2 kg
£1088.42
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

As an intermediate and in electronicsNiobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in the food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators including linear and non-linear optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1313-96-8
265.81
ZKATWMILCYLAPD-UHFFFAOYSA-N
123105
Nb2O5
MFCD00011128
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
O=[Nb](=O)O[Nb](=O)=O

Specifications

1520°C
White
99.5% (metals basis)
MFCD00011128
Insoluble in water. Soluble in HF and alkalies.
O=[Nb](=O)O[Nb](=O)=O
265.81
265.81
White powder or lump
Niobium(V) oxide
1313-96-8
Powder
Nb2O5
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
ZKATWMILCYLAPD-UHFFFAOYSA-N
dioxoniobiooxy(dioxo)niobium
123105
Odorless
4.6 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

missing translation for 'einecsNumber' : 215-213-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only