missing translation for 'onlineSavingsMsg'
Learn More
Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11347938

Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11347938
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11347938

Brand: Thermo Scientific Alfa Aesar 036478.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Vacuum Deposition

Niobium(V) oxide is used for vacuum deposition, as an alternate electrode to lithium metal in advanced fuel cells, Applied in the solid state formation of an unusual cation deficient perovskite. And it is also used in Electrical and Electronic Products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-96-8
Molecular Formula Nb2O5
Molecular Weight (g/mol) 265.81
MDL Number MFCD00011128
InChI Key ZKATWMILCYLAPD-UHFFFAOYSA-N
Synonym niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxdeShow MoreShow Less
PubChem CID 123105
SMILES O=[Nb](=O)O[Nb](=O)=O

Specifications

Melting Point 1520°C
Color White
Physical Form Sintered Lump
Quantity 100 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Soluble in HF. Insoluble in water.
IUPAC Name dioxoniobiooxy(dioxo)niobium
Formula Weight 265.81
Odor Odorless
Appearance White 3-12mm Sintered Lump
Density 4.6
Chemical Name or Material Niobium(V) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR12I027 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number036478.14,036478.22
Lot NumberY29H022 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number036478.14,036478.22
Lot NumberC12S003 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036478.14,036478.22
Lot NumberP23E037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036478.14,036478.22
Lot NumberK08Y026 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036478.14,036478.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.