Learn More
Niobium(V) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
£33.57 - £375.87
Chemical Identifiers
CAS | 1313-96-8 |
---|---|
Molecular Formula | Nb2O5 |
Molecular Weight (g/mol) | 265.81 |
MDL Number | MFCD00011128 |
InChI Key | ZKATWMILCYLAPD-UHFFFAOYSA-N |
Synonym | niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde |
PubChem CID | 123105 |
SMILES | O=[Nb](=O)O[Nb](=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10785853
|
Thermo Scientific Acros
193630100 |
10 g | Glass bottle |
£33.57
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10153970
|
Thermo Scientific Acros
193630500 |
50 g | Plastic bottle |
£108.54
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10229693
|
Thermo Scientific Acros
193632500 |
250 g | Plastic bottle |
£375.87
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
1313-96-8 | |
265.81 | |
ZKATWMILCYLAPD-UHFFFAOYSA-N | |
123105 |
Nb2O5 | |
MFCD00011128 | |
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde | |
O=[Nb](=O)O[Nb](=O)=O |
Specifications
1520.0°C | |
White | |
100.0 | |
99.99% | |
Nb2O5 | |
15, 6645 | |
Solubility in water: insoluble. Other solubilities: soluble in hf and hot h2so4, soluble in fused potassium hydrogen sulfate or, carbonates or hydroxides of the alkali metals | |
O=[Nb](=O)O[Nb](=O)=O | |
265.81 | |
265.8 | |
Trace Metal Basis | |
4.4700g/mL |
1313-96-8 | |
99.95 | |
Powder | |
Nb2O5 | |
MFCD00011128 | |
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde | |
ZKATWMILCYLAPD-UHFFFAOYSA-N | |
dioxoniobiooxy(dioxo)niobium | |
123105 | |
99.99% | |
4.47 | |
Niobium(V) oxide |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 215-213-6
RUO – Research Use Only