missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) oxide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™

Catalog Number 15403795
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 Ea.
MDL Number MFCD00011128
Formula Weight 265.81
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Niobium(V) oxide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'