missing translation for 'onlineSavingsMsg'
Learn More

Ansell Nipro GAMMEX™ Latex Surgical Undergloves

Catalog Number 17853812
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
This item is not returnable. View return policy
This product contains natural rubber latex, which may cause allergic reactions.
This item is not returnable. View return policy
This product contains natural rubber latex, which may cause allergic reactions.

These latex, powder-free surgical undergloves enable effortless double gloving and help keep the skin moist and supple in healthcare settings.

  • ASTM D6978 certified
  • Contains glycerol and dimethicone as moisturizing agents to help retain moisture and rehydrate skin
  • Made with Ansell's unique anatomic formers to enhance comfort and fit to help reduce hand fatigue
  • Green colored, smooth and with micro-textured external finish for double gloving and easy identification of a glove breach
  • DERMASHIELD™ Technology internal coating, together with the built-in moisturizer, facilitates dry and damp donning as well as intra-operative changes
  • For use in chemotherapy and healthcare applications
Certifications/Compliance ASTM D6978
Certifications/Compliance (Europe) EN 16523-1, EN 374:2003, EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016, AS/NZS 4179
Accelerator Free No
Applications Healthcare
AQL 0.65
Color Green
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 9.06 ±0.79 mil
Finger Thickness (Metric) 0.23 ±0.02 mm
Hand Specific
Laboratory/Cleanroom Use No
Latex Free No
Material Latex
Palm Thickness (English) 8.27 ±0.79 mil
Palm Thickness (Metric) 0.21 ±0.02 mm
Powdered Powder Free
Size 8.5
Sterility Sterile
Thickness (English) 8.27 ±0.79 mil
Thickness (Metric) 0.21 ±0.02 mm
Model GAMMEX-330050
Length (English) 11.4 in.
Length (Metric) 290 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'