Learn More
Reichert™ NIST Certified Calibration Oil
Check the accuracy of an instrument using calibration oils. Reichert™ NIST Certified Calibration Oil are formulated laser liquid calibration oils that are supplied with an NIST certification of traceability.
£329.00 - £367.00
Specifications
Type | NIST Certified Calibration Oil |
---|---|
For Use With (Equipment) | Refractometer |
Includes | NIST certification of traceability |
Product Code | Brand | ScaleRange | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | ScaleRange | Price | Quantity & Availability | |||||
12958564
|
Reichert™
13K41330 |
89.16 Brix |
£367.00
5oz |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16317651
|
Reichert™
13K41390 |
18.04 Brix |
£329.00
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Formulated, measured, and certified to five significant places of refractive index (0.00001).Specifications
NIST Certified Calibration Oil | |
Refractometer | |
NIST certification of traceability |