Learn More
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets
Offers the highest possible pH precision. Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets are specified to three decimal places and come with a printed certificate that guarantees traceability. Individual packets guarantee fresh solution for every calibration, as well as maximum ease of use.
£128.00 - £136.22
Specifications
For Use With (Equipment) | pH Electrode |
---|---|
For Use With (Application) | pH measurement, electrode calibration |
NIST Traceable | Yes |
Quantity | 30 x 20 mL |
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
15392265
|
Mettler Toledo™
30111136 |
4.006 (at 25°C) |
£133.00
Each |
In Stock
Log in to see stock. |
|||||
15312275
|
Mettler Toledo™
30111137 |
6.865 (at 25°C) |
£136.22
Each |
In Stock
Log in to see stock. |
|||||
15322275
|
Mettler Toledo™
30111138 |
9.180 (at 25°C) |
£128.00
Each |
In Stock
Log in to see stock. |
|||||
Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. Maximum precision is guaranteed with fully traceable NIST/DIN buffers. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
- NIST/DIN pH buffers are specified and guaranteed to three decimal places, offering top precision
- Individual sachets guarantee fresh solution for every calibration, as well as maximum ease of use
- A test certificate and detailed label with a temperature table comes with every box
- All SDS and labels contain information according to GHS (Globally Harmonized System)
Specifications
pH Electrode | |
pH measurement, electrode calibration | |
Yes | |
30 x 20 mL |